Assay Details
| Gene Symbol: LOC4345056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795237.2 | XP_015650723.1 | 7 - 8 | 1656 | 149 | |
| GenBank mRNA | AK100945.1 | - | 7 - 8 | 1604 | 149 | |
More Information
Set Membership: |
|

LOC4345056
Genomic Map