Assay Details
| Gene Symbol: LOC4345227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794549.2 | XP_015650035.1 | 3 - 4 | 789 | 106 | |
| XM_015794550.2 | XP_015650036.1 | 3 - 4 | 789 | 106 | ||
| XM_015794551.2 | XP_015650037.1 | 3 - 4 | 789 | 106 | ||
| GenBank mRNA | AK100410.1 | - | 3 - 4 | 969 | 106 | |
| AK105754.1 | - | 1 - 2 | 260 | 106 | ||
More Information
Set Membership: |
|

LOC4345227
Genomic Map