Assay Details
| Gene Symbol: LOC4345227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794549.2 | XP_015650035.1 | 4 - 5 | 908 | 99 | |
| XM_015794550.2 | XP_015650036.1 | 4 - 5 | 908 | 99 | ||
| GenBank mRNA | AK100410.1 | - | 4 - 5 | 1088 | 99 | |
| AK105754.1 | - | 2 - 3 | 379 | 99 | ||
More Information
Set Membership: |
|

LOC4345227
Genomic Map