Assay Details
| Gene Symbol: LOC4345227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794549.2 | XP_015650035.1 | 7 - 8 | 3358 | 112 | |
| XM_015794551.2 | XP_015650037.1 | 7 - 8 | 3346 | 112 | ||
| GenBank mRNA | AK100410.1 | - | 7 - 8 | 3538 | 112 | |
| AK105754.1 | - | 5 - 6 | 2828 | 112 | ||
More Information
Set Membership: |
|

LOC4345227
Genomic Map