Assay Details
| Gene Symbol: LOC4345386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792702.2 | XP_015648188.1 | 2 - 3 | 639 | 82 | |
| XM_015792703.2 | XP_015648189.1 | 2 - 3 | 639 | 82 | ||
| GenBank mRNA | AK102062.1 | - | 2 - 3 | 593 | 82 | |
More Information
Set Membership: |
|

LOC4345386
Genomic Map