Assay Details
Gene Symbol: LOC4345386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792702.2 | XP_015648188.1 | 5 - 6 | 991 | 126 | |
XM_015792703.2 | XP_015648189.1 | 5 - 6 | 991 | 126 | ||
GenBank mRNA | AK059023.1 | - | 4 - 5 | 381 | 126 | |
AK102062.1 | - | 5 - 6 | 945 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
