Assay Details
| Gene Symbol: LOC4345606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795403.2 | XP_015650889.1 | 11 - 12 | 1285 | 119 | |
| GenBank mRNA | AB096083.1 | - | 11 - 12 | 1099 | 119 | |
| AK060461.1 | - | 8 - 9 | 757 | 119 | ||
| AK071221.1 | - | 11 - 12 | 1247 | 119 | ||
| EU770323.1 | - | 11 - 12 | 1091 | 119 | ||
| EU770324.1 | - | 11 - 12 | 1092 | 119 | ||
More Information
Set Membership: |
|

LOC4345606
Genomic Map