Assay Details
| Gene Symbol: LOC4345606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795403.2 | XP_015650889.1 | 12 - 13 | 1443 | 77 | |
| GenBank mRNA | AB096083.1 | - | 12 - 13 | 1257 | 77 | |
| AK060461.1 | - | 9 - 10 | 915 | 77 | ||
| AK071221.1 | - | 12 - 13 | 1405 | 77 | ||
| DQ910546.1 | - | 12 - 13 | 1257 | 77 | ||
| EU770323.1 | - | 12 - 13 | 1249 | 77 | ||
| EU770324.1 | - | 12 - 13 | 1250 | 77 | ||
More Information
Set Membership: |
|

LOC4345606
Genomic Map