Assay Details
| Gene Symbol: LOC4345606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795403.2 | XP_015650889.1 | 13 - 14 | 1495 | 87 | |
| GenBank mRNA | AB096083.1 | - | 13 - 14 | 1309 | 87 | |
| AK060461.1 | - | 10 - 11 | 967 | 87 | ||
| AK071221.1 | - | 13 - 14 | 1457 | 87 | ||
| DQ910546.1 | - | 13 - 14 | 1309 | 87 | ||
| EU770323.1 | - | 13 - 14 | 1301 | 87 | ||
| EU770324.1 | - | 13 - 14 | 1302 | 87 | ||
More Information
Set Membership: |
|

LOC4345606
Genomic Map