Assay Details
| Gene Symbol: LOC4345606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795403.2 | XP_015650889.1 | 3 - 4 | 522 | 64 | |
| GenBank mRNA | AB096083.1 | - | 3 - 4 | 336 | 64 | |
| AK071221.1 | - | 3 - 4 | 483 | 64 | ||
| DQ910546.1 | - | 3 - 4 | 336 | 64 | ||
| EU770323.1 | - | 3 - 4 | 336 | 64 | ||
| EU770324.1 | - | 3 - 4 | 329 | 64 | ||
More Information
Set Membership: |
|

LOC4345606
Genomic Map