Assay Details
| Gene Symbol: LOC4345606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795403.2 | XP_015650889.1 | 7 - 8 | 953 | 119 | |
| GenBank mRNA | AB096083.1 | - | 7 - 8 | 767 | 119 | |
| AK060461.1 | - | 4 - 5 | 425 | 119 | ||
| AK071221.1 | - | 7 - 8 | 914 | 119 | ||
| DQ910546.1 | - | 7 - 8 | 767 | 119 | ||
| EU770324.1 | - | 7 - 8 | 760 | 119 | ||
More Information
Set Membership: |
|

LOC4345606
Genomic Map