Assay Details
| Gene Symbol: LOC4345659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795367.2 | XP_015650853.1 | 1 - 2 | 528 | 99 | |
| GenBank mRNA | AK070338.1 | - | 1 - 2 | 451 | 99 | |
More Information
Set Membership: |
|

LOC4345659
Genomic Map