Assay Details
| Gene Symbol: LOC4347314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755567.2 | XP_015611053.1 | 1 - 2 | 217 | 113 | |
| GenBank mRNA | AK102696.1 | - | 1 - 2 | 143 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755567.2 transcripts.
Set Membership: |
|

LOC4347314
Genomic Map