Assay Details
| Gene Symbol: LOC4347314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755567.2 | XP_015611053.1 | 2 - 3 | 317 | 55 | |
| GenBank mRNA | AK102696.1 | - | 2 - 3 | 243 | 55 | |
More Information
Set Membership: |
|

LOC4347314
Genomic Map