Assay Details
| Gene Symbol: LOC4348471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758554.1 | XP_015614040.1 | 6 - 7 | 1488 | 68 | |
| GenBank mRNA | AK065000.1 | - | 6 - 7 | 1378 | 68 | |
More Information
Set Membership: |
|

LOC4348471
Genomic Map