Assay Details
| Gene Symbol: LOC4325687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765414.2 | XP_015620900.1 | 1 - 2 | 373 | 122 | |
| GenBank mRNA | AK111908.1 | - | 1 - 2 | 311 | 122 | |
| AK112014.1 | - | 1 - 2 | 303 | 122 | ||
More Information
Set Membership: |
|

LOC4325687
Genomic Map