Assay Details
| Gene Symbol: LOC4323865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769654.2 | XP_015625140.1 | 19 - 20 | 3393 | 94 | |
| GenBank mRNA | AK058355.1 | - | 1 - 2 | 61 | 94 | |
| AK061474.1 | - | 1 - 2 | 61 | 94 | ||
More Information
Set Membership: |
|

LOC4323865
Genomic Map