Assay Details
| Gene Symbol: LOC4323865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769654.2 | XP_015625140.1 | 21 - 22 | 3756 | 68 | |
| GenBank mRNA | AK058355.1 | - | 2 - 3 | 502 | 68 | |
| AK061474.1 | - | 3 - 4 | 424 | 68 | ||
More Information
Set Membership: |
|

LOC4323865
Genomic Map