Assay Details
| Gene Symbol: LOC4323865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769654.2 | XP_015625140.1 | 23 - 24 | 4178 | 76 | |
| GenBank mRNA | AK058355.1 | - | 4 - 5 | 924 | 76 | |
| AK061474.1 | - | 5 - 6 | 846 | 76 | ||
More Information
Set Membership: |
|

LOC4323865
Genomic Map