Assay Details
| Gene Symbol: LOC4325448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765394.2 | XP_015620880.1 | 7 - 8 | 1295 | 79 | |
| GenBank mRNA | AK071525.1 | - | 7 - 8 | 1252 | 79 | |
More Information
Set Membership: |
|

LOC4325448
Genomic Map