Assay Details
| Gene Symbol: LOC4324049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766895.2 | XP_015622381.1 | 3 - 4 | 663 | 152 | |
| XM_015766896.2 | XP_015622382.1 | 3 - 4 | 664 | 152 | ||
| GenBank mRNA | AK102473.1 | - | 3 - 4 | 647 | 152 | |
More Information
Set Membership: |
|

LOC4324049
Genomic Map