Assay Details
| Gene Symbol: LOC4324791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773654.2 | XP_015629140.1 | 1 | 1032 | 134 | |
| GenBank mRNA | AK068115.1 | - | 1 | 854 | 134 | |
More Information
Set Membership: |
|

LOC4324791
Genomic Map