Assay Details
| Gene Symbol: LOC4324367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766370.2 | XP_015621856.1 | 2 - 3 | 260 | 67 | |
| GenBank mRNA | AK059028.1 | - | 2 - 3 | 203 | 67 | |
More Information
Set Membership: |
|

LOC4324367
Genomic Map