Assay Details
| Gene Symbol: LOC4325383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769945.2 | 1 - 2 | 992 | 102 | ||
| GenBank mRNA | AK059235.1 | - | 1 - 2 | 247 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325383| Gene Symbol: LOC4325383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769945.2 | 1 - 2 | 992 | 102 | ||
| GenBank mRNA | AK059235.1 | - | 1 - 2 | 247 | 102 | |
Set Membership: |
|