Assay Details
| Gene Symbol: LOC4345812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795212.1 | XP_015650698.1 | 1 - 2 | 216 | 62 | |
| GenBank mRNA | AK064071.1 | - | 1 - 2 | 212 | 62 | |
| AK064300.1 | - | 1 - 2 | 213 | 62 | ||
| AK103413.1 | - | 1 - 2 | 216 | 62 | ||
| AK106526.1 | - | 1 - 2 | 212 | 62 | ||
| EU837245.1 | - | 1 - 2 | 191 | 62 | ||
More Information
Set Membership: |
|

LOC4345812
Genomic Map