Assay Details
| Gene Symbol: LOC9271198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793313.2 | XP_015648799.1 | 3 - 4 | 748 | 88 | |
| XM_015793314.2 | XP_015648800.1 | 3 - 4 | 748 | 88 | ||
| GenBank mRNA | AK242582.1 | - | 3 - 4 | 780 | 88 | |
More Information
Set Membership: |
|

LOC9271198
Genomic Map