Assay Details
| Gene Symbol: LOC4345907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794584.2 | XP_015650070.1 | 1 - 2 | 904 | 68 | |
| XM_015794586.2 | XP_015650072.1 | 2 - 3 | 799 | 68 | ||
| GenBank mRNA | AK103656.1 | - | 1 - 2 | 268 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794584.2, XM_015794586.2 transcripts.
Set Membership: |
|

LOC4345907
Genomic Map