Assay Details
| Gene Symbol: LOC4345926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792794.2 | XP_015648280.1 | 1 - 2 | 374 | 76 | |
| GenBank mRNA | AK103187.1 | - | 1 - 2 | 338 | 76 | |
| AK120031.1 | - | 1088 | 76 | |||
More Information
Set Membership: |
|

LOC4345926
Genomic Map