Assay Details
| Gene Symbol: LOC9271005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793895.2 | XP_015649381.1 | 18 - 19 | 2396 | 83 | |
| GenBank mRNA | AK060581.1 | - | 6 - 7 | 737 | 83 | |
| AK071334.1 | - | 16 - 17 | 2657 | 83 | ||
More Information
Set Membership: |
|

LOC9271005
Genomic Map