Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 11 - 12 | 1976 | 72 | |
| GenBank mRNA | AK067738.1 | - | 10 - 11 | 1335 | 72 | |
| AK100974.1 | - | 11 - 12 | 1977 | 72 | ||
| AY224568.1 | - | 9 - 10 | 1148 | 72 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map