Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 15 - 16 | 2409 | 127 | |
| XM_015756767.2 | XP_015612253.1 | 15 - 16 | 2487 | 127 | ||
| GenBank mRNA | AK058665.1 | - | 5 - 6 | 505 | 127 | |
| AK067738.1 | - | 14 - 15 | 1768 | 127 | ||
| AK100974.1 | - | 15 - 16 | 2410 | 127 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map