Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 17 - 18 | 2563 | 68 | |
| XM_015756767.2 | XP_015612253.1 | 17 - 18 | 2641 | 68 | ||
| GenBank mRNA | AK058665.1 | - | 7 - 8 | 659 | 68 | |
| AK067738.1 | - | 16 - 17 | 1922 | 68 | ||
| AK100974.1 | - | 17 - 18 | 2564 | 68 | ||
| AY224568.1 | - | 15 - 16 | 1747 | 68 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map