Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 19 - 20 | 2681 | 90 | |
| XM_015756767.2 | XP_015612253.1 | 19 - 20 | 2759 | 90 | ||
| GenBank mRNA | AK067738.1 | - | 18 - 19 | 2040 | 90 | |
| AK100974.1 | - | 19 - 20 | 2682 | 90 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map