Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 21 - 22 | 2879 | 68 | |
| XM_015756767.2 | XP_015612253.1 | 21 - 22 | 2957 | 68 | ||
| GenBank mRNA | AK058665.1 | - | 11 - 12 | 975 | 68 | |
| AK067738.1 | - | 20 - 21 | 2238 | 68 | ||
| AK100974.1 | - | 21 - 22 | 2880 | 68 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map