Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 22 - 23 | 3099 | 123 | |
| XM_015756767.2 | XP_015612253.1 | 22 - 23 | 3177 | 123 | ||
| GenBank mRNA | AK058665.1 | - | 12 - 13 | 1195 | 123 | |
| AK067738.1 | - | 21 - 22 | 2458 | 123 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map