Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 2 - 3 | 808 | 76 | |
| XM_015756767.2 | XP_015612253.1 | 2 - 3 | 889 | 76 | ||
| GenBank mRNA | AK067738.1 | - | 1 - 2 | 167 | 76 | |
| AK100974.1 | - | 2 - 3 | 809 | 76 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map