Assay Details
| Gene Symbol: LOC4346402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756766.2 | XP_015612252.1 | 9 - 10 | 1771 | 92 | |
| XM_015756767.2 | XP_015612253.1 | 9 - 10 | 1852 | 92 | ||
| GenBank mRNA | AK067738.1 | - | 8 - 9 | 1130 | 92 | |
| AK100974.1 | - | 9 - 10 | 1772 | 92 | ||
| AY224568.1 | - | 7 - 8 | 943 | 92 | ||
More Information
Set Membership: |
|

LOC4346402
Genomic Map