Assay Details
| Gene Symbol: LOC4346539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757069.2 | XP_015612555.1 | 2 - 3 | 1266 | 78 | |
| GenBank mRNA | AK070835.1 | - | 2 - 3 | 1267 | 78 | |
More Information
Set Membership: |
|

LOC4346539
Genomic Map