Assay Details
| Gene Symbol: LOC4346606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757207.2 | XP_015612693.1 | 6 - 7 | 704 | 90 | |
| XM_015757209.2 | XP_015612695.1 | 6 - 7 | 706 | 90 | ||
| GenBank mRNA | AK101452.1 | - | 6 - 7 | 688 | 90 | |
| AY371050.1 | - | 6 - 7 | 648 | 90 | ||
More Information
Set Membership: |
|

LOC4346606
Genomic Map