Assay Details
| Gene Symbol: LOC4346606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757207.2 | XP_015612693.1 | 9 - 10 | 986 | 80 | |
| XM_015757209.2 | XP_015612695.1 | 9 - 10 | 988 | 80 | ||
| GenBank mRNA | AK101452.1 | - | 9 - 10 | 970 | 80 | |
| AY371050.1 | - | 9 - 10 | 930 | 80 | ||
More Information
Set Membership: |
|

LOC4346606
Genomic Map