Assay Details
| Gene Symbol: LOC4347373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756674.1 | XP_015612160.1 | 1 - 2 | 334 | 104 | |
| GenBank mRNA | AK100536.1 | - | 1 - 2 | 340 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756674.1 transcripts.
Set Membership: |
|

LOC4347373
Genomic Map