Assay Details
| Gene Symbol: LOC9270778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756481.2 | XP_015611967.1 | 6 - 7 | 1156 | 116 | |
| XM_015756482.2 | XP_015611968.1 | 5 - 6 | 1132 | 116 | ||
| GenBank mRNA | AB510402.1 | - | 5 - 6 | 1110 | 116 | |
| AK061724.1 | - | 2 - 3 | 305 | 116 | ||
| AK065118.1 | - | 4 - 5 | 1225 | 116 | ||
More Information
Set Membership: |
|

LOC9270778
Genomic Map