Assay Details
| Gene Symbol: LOC9270778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756481.2 | XP_015611967.1 | 8 - 9 | 1402 | 150 | |
| XM_015756482.2 | XP_015611968.1 | 7 - 8 | 1378 | 150 | ||
| GenBank mRNA | AB510402.1 | - | 7 - 8 | 1356 | 150 | |
| AK061724.1 | - | 4 - 5 | 551 | 150 | ||
| AK065118.1 | - | 6 - 7 | 1471 | 150 | ||
More Information
Set Membership: |
|

LOC9270778
Genomic Map