Assay Details
| Gene Symbol: LOC4347749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756133.2 | XP_015611619.1 | 7 - 8 | 986 | 118 | |
| GenBank mRNA | AK072690.1 | - | 7 - 8 | 862 | 118 | |
More Information
Set Membership: |
|

LOC4347749
Genomic Map