Assay Details
| Gene Symbol: LOC4347752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795661.2 | XP_015651147.1 | 3 - 4 | 326 | 91 | |
| XM_015795662.2 | XP_015651148.1 | 3 - 4 | 327 | 91 | ||
| GenBank mRNA | AK071684.1 | - | 3 - 4 | 289 | 91 | |
More Information
Set Membership: |
|

LOC4347752
Genomic Map