Assay Details
| Gene Symbol: LOC4347752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795661.2 | XP_015651147.1 | 5 - 6 | 474 | 92 | |
| XM_015795662.2 | XP_015651148.1 | 5 - 6 | 475 | 92 | ||
| GenBank mRNA | AK071684.1 | - | 5 - 6 | 437 | 92 | |
More Information
Set Membership: |
|

LOC4347752
Genomic Map