Assay Details
| Gene Symbol: LOC4348081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758484.2 | XP_015613970.1 | 1 - 2 | 324 | 60 | |
| GenBank mRNA | AK100609.1 | - | 1 - 2 | 304 | 60 | |
More Information
Set Membership: |
|

LOC4348081
Genomic Map