Assay Details
| Gene Symbol: LOC4348081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758484.2 | XP_015613970.1 | 2 - 3 | 920 | 61 | |
| GenBank mRNA | AK100609.1 | - | 2 - 3 | 900 | 61 | |
More Information
Set Membership: |
|

LOC4348081
Genomic Map