Assay Details
| Gene Symbol: LOC4348150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757740.2 | XP_015613226.1 | 1 - 2 | 67 | 68 | |
| GenBank mRNA | AK064267.1 | - | 1 - 2 | 58 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757740.2 transcripts.
Set Membership: |
|

LOC4348150
Genomic Map