Assay Details
| Gene Symbol: LOC4348150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757740.2 | XP_015613226.1 | 2 - 3 | 309 | 53 | |
| GenBank mRNA | AK064267.1 | - | 2 - 3 | 300 | 53 | |
More Information
Set Membership: |
|

LOC4348150
Genomic Map